Learn More
SLC12A6 Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142299
Additional Details : Weight : 0.01000kg
Description
Additional Information: This antibody is expected to recognise all reported isofoms (NP_598408.1; NP_005126.1; NP_001035959.1; NP_001035961.1; NP_001035962.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
SLC12A6 is a member of the K-Cl cotransporter (KCC) family. K-Cl cotransporters are integral membrane proteins that lower intracellular chloride concentrations below the electrochemical equilibrium potential. The proteins encoded by this gene are activated by cell swelling induced by hypotonic conditions. Alternate splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are associated with agenesis of the corpus callosum with peripheral neuropathy.Specifications
SLC12A6 | |
Polyclonal | |
Biotin | |
SLC12A6 | |
9530023I19Rik; ACCPN; DKFZP434D2135; Electroneutral potassium-chloride cotransporter 3; furosemide-sensitive KCl cotransporter 3; gaxp; Kcc3; KCC3A; KCC3B; K-Cl cotransporter 3; potassium chloride cotransporter 3; potassium chloride cotransporter KCC3a-S3; potassium-chloride transporter-3a; potassium-chloride transporter-3b; SLC12A6; solute carrier family 12 (potassium/chloride transporter), member 6; solute carrier family 12 (potassium/chloride transporters), member 6; solute carrier family 12 member 6; solute carrier family 12, member 6 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
9990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UHW9 | |
SLC12A6 | |
Peptide with sequence SRHIDVCSKTKE. | |
100 μg | |
Primary | |
Human | |
Liquid |