missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SKIV2L Monoclonal Antibody (K94011_10D9), Invitrogen™

Product Code. p-200088743
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30210252 200 μL 200µg
30194411 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30210252 Supplier Invitrogen™ Supplier No. MA556092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: SEFPSRKDSK AHEQALAELT KRLGALEEPD MTGQLVDLPE YYSWGEELTE TQHMIQRRIM ESVNGLKSLS AGRVVVVKNQ EHHNALGVIL QVSSNSTSRV FTTLVLCDKP LSQDPQDRGP ATAEVPYPDD LVGFKLFLPE GPCDHTVVKL QPGDMAAITT KVLRVNGEKI LEDFSKRQQP KFKKDPPLAA VTTAVQELLR LAQAHPAGPP TLDPVNDLQL KDMSVVEGGL RARKLEELIQ GAQCVHSPRF PAQYLKLRER MQIQKEMERL

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a human homologue of yeast SKI2 and may be involved in antiviral activity by blocking translation of poly(A) deficient mRNAs. This gene is located in the class III region of the major histocompatibility complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKIV2L
Applications Western Blot
Classification Monoclonal
Clone K94011_10D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SKIV2L
Gene Accession No. Q15477
Gene Alias 170A; 4930534J06Rik; AW214248; Ddx13; Helicase SKI2W; helicase-like protein; HLP; SKI; SKI2; SKI2 homolog, superkiller viralicidic activity 2-like; Ski2 like RNA helicase; Ski2w; SKIV2; SKIV2L; SKIV2L1; superkiller viralicidic activity 2-like; superkiller viralicidic activity 2-like (S. cerevisiae); THES2; W
Gene Symbols SKIV2L
Host Species Mouse
Immunogen Recombinant human SKIC2 (immunogen range is 771-1040).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6499
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.