Learn More
Honeywell™ SKELETON NIT 5 Gloves
Click to view available options
Size:
8
9
10
Unit Size:
1 pair
Description
- Knitted glove in composite and para-aramid yarn
- The knitting technique provides the glove with excellent fit and high dexterity
- The composite yarn is reliable and safe protection
- The para-aramid yarn assure a resistance to the contact heat and cut risks
- Plated polyamide inside
- Offer a greater comfort and limit the contact of the skin with composite fiber
- Black nitrile foam coating on palm
- Nitrile coating provides good protection against oil and grease
- The foam coating makes the glove more flexible than standard nitrile
- Rubber pad on top of the hand and fingers
- Helps to protect metacarpian and fingers from shocks
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards |
| Description | 13 gauge |
| Color | Yellow, Green |
| Material | Nitrile, Polyamide |
| Size | 10 |
| Length (Metric) | 25 cm |