missing translation for 'onlineSavingsMsg'
Learn More

SKA-31, MedChemExpress

Product Code. p-200096810
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16467227

Brand: Medchem Express HY111655

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKA-31 is a potent potassium channel activator with EC50s of 260 nM, 1.9 μM, 2.9 μM, and 2.9 μM for KCa3.1, KCa2.2, KCa2.1 and KCa2.3, respectively. SKA-31 potentiates endothelium-derived hyperpolarizing factor response and lowers blood pressure.

SKA-31 is a potent potassium channel activator with EC50s of 260 nM, 1.9 μM, 2.9 μM, and 2.9 μM for KCa3.1, KCa2.2, KCa2.1 and KCa2.3, respectively. SKA-31 potentiates endothelium-derived hyperpolarizing factor response and lowers blood pressure.

Specifications

Chemical Name or Material SKA-31
Physical Form Solid
CAS 40172-65-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H8N2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 150 mg/mL (749.03 mM; Need ultrasonic)
SMILES NC1=NC2=C3C=CC=CC3=CC=C2S1
Molecular Weight (g/mol) 200.26
Formula Weight 200.26
Percent Purity 99.45%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.