Learn More
Masterflex™ Six Channel Syringe Pump
Product Code. 11759889
Change view
Click to view available options
Flow Rate:
22.3 mL/min.
Unit Size:
Each
Product Code. Flow Rate unitSize
11759889 22.3 mL/min. Each
1 options
Product Code. 11759889
Brand: Masterflex™ 9002436
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Six-channel pump can both withdraw and infuse
  • Programming features:
    • Programming commands allow dispensing of a specified amount, automatic rates changes, direction changes, programmed pauses, external or user input, and sending or receive signals from other equipment.
    • Non-volatile memory restores all setup parameters and the pumping program upon turning the pump on.
    • An audible buzzer can be programmed to alert when an alarm condition occurs or the pumping program completes.
    • Syringe purge mode will infuse or refill syringe at top speed to purge air from the line or to fill the syringe.
    • Versatile pumping features allow infusion or withdrawal in μl/hr, μl/min, ml/hr, and mL/min.
  • Communication features:
    • Network up to 100 pumps from a single communications port on a computer.
    • Network communication protocols allow the ability to send a network burst transmission simultaneously to 10 pumps in the network with no lag, resulting in consistent laboratory setups.
    • Optional communications protocol utilizes error checking to ensure completely reliable signal transmissions by eliminating errors caused by disconnections or electromagnetic interference.
Specifications
Voltage 230 V
Flow Rate 22.3 mL/min.
Capacity (Metric) 6 x 60 mL
Dimensions (L x W x H) Exterior 15.25 in. X 10.5 in. X 4.75 in.
Number of Channels 6