Learn More
Sisomicin sulfate, MedChemExpress
Description
Sisomicin is a broad-spectrum aminoglycoside antibiotic produced by Micromonospora inyoensis. sisomicin has great activity against gram-positive bacteria.
Specifications
Specifications
| Chemical Name or Material | Sisomicin sulfate |
| CAS | 53179-09-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H37N5O7.5/2H2SO4 |
| Quantity | 250 mg |
| Solubility Information | H2O : 20 mg/mL (28.87 mM; Need ultrasonic and warming) |
| SMILES | N[C@H]1[C@](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@@]3([H])OC[C@@](O)(C)[C@H](NC)[C@H]3O)[C@H]2O)([H])OC(CN)=CC1.[2.5H2SO4] |
| Molecular Weight (g/mol) | 692.72 |
| Formula Weight | 692.72 |
| Show More |