missing translation for 'onlineSavingsMsg'
Learn More

SIRT7 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 17689686 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17689686 20 μL 20µL
17679686 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17689686

Brand: Proteintech 297291AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SIRT7 Polyclonal antibody apecifically detetects SIRT7 antigen in Human Samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SIRT7
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 800 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. NM_016538
Gene Symbols SIRT7
Host Species Rabbit
Immunogen SIRT7 fusion protein Ag31794
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 51547
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.