Learn More
SIRT6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13572-1-AP-150UL
 View more versions of this product
Product Code. 16851854
  • 4836.10 NOK / 150µL
Description
This antibody is a rabbit polyclonal antibody raised against full length human SIRT6 antigen.
The Silent Information Regulator (SIR2) family of genes are highly conserved from prokaryotes to eukaryotes and have important functions in the regulation of metabolism, growth and differentiation, inflammation, cellular survival, as well as in senescence and lifespan extension. Sirtuins, including SIRT1-7, are human homologs of yeast Sir2p. Sirtuins are NAD+-dependent histone/protein deacetylases (HDAC) which regulate cellular metabolism, e.g. energy metabolism, and thereby are associated with aging and several age-related diseases. SIRT6 is a histone H3 lysine 9 deacetylase and is thought to stabilize the DNA-dependent protein kinase at double-stranded DNA breaks and may have a role in the process of mammalian aging.
Specifications
Specifications
SIRT6
Polyclonal
Unconjugated
SIRT6
SIR2 like protein 6, SIR2L6, SIRT6
Rabbit
Antigen Affinity Chromatography
RUO
299638, 50721, 51548
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59941, Q8N6T7
SIRT6
SIRT6 Fusion Protein Ag4469
150 μL
Primary
Human, Mouse, Rat