Learn More
SIRT5 inhibitor 1, MedChemExpress
Product Code. 16497377
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16497377
Brand: Medchem Express HY112634
MedChemExpress SIRT5 inhibitor 1 is a potent Human Sirtuin 5 deacylase inhibitor, with an IC50 of 0.11 μM.
SIRT5 inhibitor 1 is a potent Human Sirtuin 5 deacylase inhibitor, with an IC50 of 0.11 μM.
Specifications
Chemical Name or Material SIRT5 inhibitor 1
CAS 2166487-21-2
Color White
Purity Grade Notes Research
Molecular Formula C31H39FN6O6S2
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (185.24 mM; Need ultrasonic)
SMILES O=S(N[C@@H](CCCCNC(NCCC(O)=O)=S)C(N[C@H](C(NC1CCC1)=O)CC2=CNC3=CC=CC=C23)=O)(C4=CC(F)=CC=C4)=O
Molecular Weight (g/mol) 674.81
Formula Weight 674.81
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.