missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29838-1-AP-150UL
This item is not returnable.
View return policy
Description
SIRT3 Polyclonal antibody specifically detects SIRT3 in Human samples. It is validated for WB, ELISASpecifications
| SIRT3 | |
| Polyclonal | |
| Unconjugated | |
| NM_016252 | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, ELISA | |
| 600 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Rabbit | |
| 150 μL | |
| 23410 | |
| Store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction