Learn More
Sinomenine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Sinomenine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Sinomenine hydrochloride |
| CAS | 6080-33-7 |
| Analyte or Component Names | Sinomenine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H23 N O4 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Sinomenine hydrochloride (6CI,7CI), Cucoline, hydrochloride, NSC 76021, Sinomenin hydrochloride, 9alpha,13alpha,14alpha-Morphinan-6-one, 7,8-didehydro-4-hydroxy-3,7-dimethoxy-17-methyl-, hydrochloride (8CI), Morphinan-6-one, 7,8-didehydro-4-hydroxy-3,7-dimethoxy-17-methyl-, hydrochloride, (9alpha,13alpha,14alpha)- (9CI), Morphinan-6-one, 7,8-didehydro-4-hydroxy-3,7-dimethoxy-17-methyl-, hydrochloride (1:1), (9alpha,13alpha,14alpha)- |
| InChI Formula | InChI=1S/C19H23NO4.ClH/c1-20-7-6-19-10-14(21)16(24-3)9-12(19)13(20)8-11-4-5-15(23-2)18(22)17(11)19;/h4-5,9,12-13,22H,6-8,10H2,1-3H3;1H/t12-,13+,19-;/m1./s1 |
| SMILES | Cl.COC1=C[C@@H]2[C@@H]3Cc4ccc(OC)c(O)c4[C@]2(CCN3C)CC1=O |
| Molecular Weight (g/mol) | 365.85 |
| Show More |