Learn More
Poulten Graf™ Single Use Luer-Lock Two-Part Syringe
Click to view available options
Unit Size:
Pack of 30
Description
- Produced in accordance with ISO 7886, ISO 7864 and ISO 8537
- PVC-free and non-toxic
- Sterilixed with ethylene oxide according to DIN 13098 and dispatched in blister packs
- Extremely tight with a transparent, smooth operating cylinder
- Screen printed graduation
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | CE |
| Material | Polypropylene |
| Description | Screen printed graduation |
| Product Type | Luer-lock 2-part Syringe |
| Disposable | Single-use |
| Quantity | 30/Pk. |
| Volume (Metric) | 50 to 60 mL |
| Connection Type | Luer Lock |
| Graduated | Yes |
| Show More |