Learn More
Thermo Scientific™ Single-Use BioProcess Container, 2D, Aegis™ 5-14, 2 ports
GREENER_CHOICE
Product Code. 17839766 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
3 L
Unit Size:
Pack of 10
Product Code. Capacity (Metric) unitSize
17839766 500 mL Pack of 10
17899756 50 mL Pack of 10
17809696 100 mL Pack of 10
17829766 250 mL Pack of 10
17899686 1 L Pack of 10
17849776 2 L Pack of 10
17859776 3 L Pack of 10
7 options
Product Code. 17839766
Brand: Thermo Scientific™ SH31265.14
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
Features:
• All BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Flexible and economic solution to update or increase your bioreactor capacity
• Suitable for both suspension and adherent cultures
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from a large component library – with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and gas filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Quantity 500 mL
Tubing Style TPE