Learn More
Masterflex Single Head Piston Pump, 10 mL/min, PEEK fluid path, 100-240 VAC
Brand:  Masterflex 74931-00
Single Head Piston Pump, 10 mL/min, PEEK fluid path, 100-240 VAC
Product Code. 15579613
  • 59300.00 NOK / Each
Only null left
Estimated Shipment: 04-03-2026