Learn More
Socorex™ Acura™ Single-channel Micropipette Volumetric Module
Description
- One of 27 available volumetric assemblies.
- Increase versatility and save costs.
- Disassembles and reassembles without tools.
- Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Specifications
| For Use With | Acura™ Electro™ 926 MicroPipette |
| Number of Channels | 1 |
| Product Line | Acura |
| Type | Electronic Pipette Volumetric Module |
| Volume (Metric) | 2.5 to 50 μL |
| For Use With (Equipment) | Acura™ Electro 926 Micropipette |
| Quantity | 1 Each |