Learn More
Socorex™ Acura™ Single-channel Micropipette Volumetric Module
Product Code. 11743276
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 200 μL
2.5 to 50 μL
50 to 1000 μL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11743276 0.5 to 10 μL Each
11773276 50 to 1000 μL Each
11753276 2.5 to 50 μL Each
11763276 10 to 200 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11743276 Supplier Socorex™ Supplier No. 800.0010
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ Single-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your single-channel micropipetting needs.
  • One of 27 available volumetric assemblies.
    • Increase versatility and save costs.
  • Disassembles and reassembles without tools.
  • Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
For Use With Acura™ Electro™ 926 MicroPipette
Number of Channels 1
Product Line Acura
Type Electronic Pipette Volumetric Module
Volume (Metric) 0.5 to 10 μL
For Use With (Equipment) Acura™ Electro 926 Micropipette
Quantity 1 Each