Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Product Code. 11881762 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
4-position carousel base
GENESYS 140/150/180,BioMate160
GENESYS 30/40/50
Spectronic200,GENESYS 30/40/50
Type:
Beckman-style Cell Holder
Cylindrical Cell Holder
Disposable Microcell Holder
Hose Entry/Exit Kit
Indiv Adjustable Filter Holder
Long Path Cell Holder
Sample Holder for Film/Filter
Single Cell Sample Holder
Test Tube Cap
Test Tube and Vial Holder
Holds:
10 mm cuvette (1)
10 to 100 mm cuvette (1)
12 mm to 25 mm tubes and vials
20 to 100 mm cuvette (1)
5 - 100 mm cylindrical cells
5 to 100 mm cuvette (1)
5 to 100mm cylindrical cuvette
50 μL microcells, Uvettes
Beckman-style short microcells
Entry and exit hoses (incl)
Filter/lenses, 10 mm thick
Filter/lenses, 6 mm thick
Filters up to 10 mm thick
Single-cell cuvette, 10mm tube
Square 10 mm PL Cuvette
Tall tubes, vials (lid open)
Unit Size:
Each
Product Code. For Use With (Equipment) Type Holds unitSize
11881762 Spectronic200,GENESYS 30/40/50 Single Cell Sample Holder Square 10 mm PL Cuvette Each
15886085 GENESYS 140/150/180,BioMate160 Single Cell Sample Holder 10 mm cuvette (1) Each
15896085 GENESYS 140/150/180,BioMate160 Beckman-style Cell Holder Beckman-style short microcells Each
15816095 GENESYS 140/150/180,BioMate160 Long Path Cell Holder 5 to 100mm cylindrical cuvette Each
15826095 GENESYS 140/150/180,BioMate160 Sample Holder for Film/Filter Filter/lenses, 10 mm thick Each
15836095 GENESYS 140/150/180,BioMate160 Disposable Microcell Holder 50 μL microcells, Uvettes Each
15866095 GENESYS 140/150/180,BioMate160 Long Path Cell Holder 10 to 100 mm cuvette (1) Each
11893744 GENESYS 30/40/50 Indiv Adjustable Filter Holder Filters up to 10 mm thick Each
15520592 GENESYS 30/40/50 Test Tube and Vial Holder 12 mm to 25 mm tubes and vials Each
15530592 GENESYS 30/40/50 Test Tube Cap Tall tubes, vials (lid open) Each
15540592 GENESYS 30/40/50 Long Path Cell Holder 20 to 100 mm cuvette (1) Each
15550592 GENESYS 30/40/50 Long Path Cell Holder 5 to 100 mm cuvette (1) Each
15570592 GENESYS 30/40/50 Sample Holder for Film/Filter Filter/lenses, 6 mm thick Each
15580592 GENESYS 30/40/50 Hose Entry/Exit Kit Entry and exit hoses (incl) Each
11751151 4-position carousel base Cylindrical Cell Holder 5 - 100 mm cylindrical cells Each
11771151 GENESYS 30/40/50 Single Cell Sample Holder Single-cell cuvette, 10mm tube Each
15520602 GENESYS 30/40/50 Single Cell Sample Holder 10 mm cuvette (1) Each
17 options
Product Code. 11881762
Brand: Thermo Scientific™ 335079000
Simplify sampling with easy access
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
TRUSTED_SUSTAINABILITY
Specifications
Description Thermostatted rectangular cell holder
Type Single Cell Sample Holder
For Use With (Equipment) Spectronic200,GENESYS 30/40/50
Holds Square 10 mm PL Cuvette
Use Use with Accessory kit (840-278200)