Learn More
Biochrom Single Cell Holder
Single cell holder for Ultrospec 2100 pro spectrophotometer.
3275.73 NOK
Product Code | Brand | Pathlength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pathlength | Price | Quantity & Availability | |||||
10680951
|
Biochrom
80-2106-05 |
10 mm |
3275.73 NOK
Each |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Temperature range from ambient to 100°C
- For use with Ultrospec 2100 pro
Specifications
Cell Holder |