Learn More
Simport™ Scientific Stainless-Steel Base Molds
For applications in specimen embedding with Simport embedding rings and cassettes
Description
- Made of Stainless Steel
- Highly polished surface
- Superior thermal exchange
- Well corners are rounded for optimal paraffin ribboning
For specimen embedding