Learn More
Simport™ Scientific EasyDip™ Slide Staining Jars
User-friendly approach for staining microscope slides. These jars are a part of the EasyDip™ Slide Staining System. (See also the EasyDip Slide Staining Rack.)
Spezifikation
| Material | Acetal Plastic |
|---|---|
| For Use With (Application) | Special stains, frozen sections and special processes |
| Height (English) | 3.62 in. |
| Height (Metric) | 92 mm |
| Slide Count | 12 standard microscope slides |
Beschreibung
- Vertical slide staining rack (sold separately) will hold up to 12 standard microscope slides
- Slide writing area is kept out of staining fluid
- Jars can be loosely joined to each other
- Made of acetal plastic that resists attacks by most staining agents (except phenol, iodine or ferric chloride)
Spezifikation
| Acetal Plastic | |
| 3.62 in. | |
| 12 standard microscope slides |
| Special stains, frozen sections and special processes | |
| 92 mm | |
| 80 mL |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts