Learn More
Silybin A,B (mixture), TRC
Specifications
Specifications
| Chemical Name or Material | Silybin (A+B) |
| CAS | 802918-57-6 |
| Molecular Formula | C25 H22 O10 |
| Quantity | 2.5 g |
| Synonym | 4H-1-Benzopyran-4-one','2-[2,3-dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-1,4-benzodioxin-6-yl]-2,3-dihydro-3,5,7-trihydroxy-','(2R,3R)-, 4H-1-Benzopyran-4-one','2-[2,3-dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-1,4-benzodioxin-6-yl]-2,3-dihydro-3,5,7-trihydroxy- (9CI), (2R,3R)-2-[2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-1,4-benzodioxin-6-yl]-2,3-dihydro-3,5,7-trihydroxy-4H-1-benzopyran-4-one, Silybin |
| InChI Formula | InChI=1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-16-5-3-12(7-18(16)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3/t20?,23-,24?,25+/m0/s1 |
| SMILES | COc1cc(ccc1O)C2Oc3cc(ccc3OC2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@@H]4O |
| IUPAC Name | (2R,3R)-3,5,7-trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydro-1,4-benzodioxin-6-yl]-2,3-dihydrochromen-4-one |
| Molecular Weight (g/mol) | 482.44 |
| Formula Weight | 482.1213 |