Learn More
Silver tetrafluoroborate, 99%
Product Code. 15740829 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Plastic bottle
Unit Size:
10g
1g
50g
Product Code. 15740829
Brand: Thermo Scientific Acros 197090500
Request Bulk or Custom Format
CAS: 14104-20-2 | AgBF4 | 194.67 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14104-20-2
Molecular Formula AgBF4
Molecular Weight (g/mol) 194.67
MDL Number MFCD00003408
InChI Key CCAVYRRHZLAMDJ-UHFFFAOYSA-N
Synonym silver tetrafluoroborate, silver borofluoride, silver fluoborate, silver fluoroborate, silver i tetrafluoroborate, borate 1-, tetrafluoro-, silver 1+, silver tetrafluroborate, acmc-209clm
PubChem CID 159722
SMILES [Ag+].F[B-](F)(F)F
Specifications
Color Beige
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 43.92% min. (BF4)
Linear Formula AgBF4
Solubility Information Solubility in water: soluble.
IUPAC Name silver(1+) tetrafluoroboranuide
Formula Weight 194.67
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Silver tetrafluoroborate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Silver tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Silver tetrafluoroborate