Learn More
Silver Chloride, 99%+, MilliporeSigma™
Product Code. 15895900
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895900 25 g Plastic bottle 25g
15805910 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15895900 Supplier Supelco Supplier No. 1192030025
 Restricted Item
Estimated Shipment: 26-11-2026
Chemical Identifiers
CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]
Specifications
Melting Point 455°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%+
Synonym Silver(1+) chloride
Formula Weight 143.32
Odor Odorless
Packaging Plastic bottle
Appearance White powder
Chemical Name or Material Silver chloride