Description
The gray coloring on the outside of our uvex silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. The inside however is pure white. In addition, the 3D nose geometry for all products in this family ensures a secure fit, even for different facial shapes.
- Particle-filtering flat-fold mask with climate-optimised exhalation valve
- Hydrophobic material coating
- Four-point headband attachment for a comfortable, individual and secure fit
- Flexible, adjustable integrated nose clip
- Very soft, all-around sealing lip for optimal wearer comfort and secure fit
- Individually sealed
- Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
- FFP3 respirator certified in accordance with EN 149:2001+A1:2009
- D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
Specifications
Specifications
| Aerosol Type | P - Oil-Proof |
| Color | White |
| Design Type | Flat-fold |
| Disposable | Reusable |
| Filter Type | FFP3 |
| Material | Polypropylene, Polyester |
| Product Type | Particulate Respirator |
| Size | Medium |
| With Exhalation Valve | Yes |
| Latex Free | Yes |
| Show More |