Learn More
Trajan™ SilTite™ FingerTite Ferrule System Adapters
For gas chromatography systems
965.00 NOK - 995.00 NOK
Product Code | Brand | Description | For Use With | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With | Price | Quantity & Availability | ||||
16602642
|
Trajan™
0736101 |
Agilent Capillary Adaptor | Agilent Capillary |
965.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15208953
|
Trajan™
0736102 |
Agilent MS Adaptor | Agilent GCs |
995.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15228953
|
Trajan™
0736104 |
Agilent Injector Adaptor | Agilent GCs, , Thermo 1300 Injector |
995.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
16612642
|
Trajan™
0736103 |
Agilent FID adapter | Agilent FID |
995.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Adapter |