Learn More
Thermo Scientific™ SilTite™ Ferrules
Product Code. 12018049 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12018049 0.25 mm Pack of 10
10025844 0.32 mm Pack of 10
11560964 0.53 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12018049 Supplier Thermo Scientific™ Supplier No. 290MF229
Estimated Shipment: 08-01-2027
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Description For 0.25 mm I.D. Capillary; Pk. of 10
Diameter (Metric) 0.25 mm
For Use With (Equipment) Fused Silica Capillary GC Columns
Material SilTite Metal