Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Product Code. p-4522208 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo Scientific DSQ GC-MS Interface
Thermo Scientific ISQ GC-MS Interface
Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Diameter (Metric) Inside:
0.1 to 0.25 mm
0.32 mm
0.53 mm
Unit Size:
Pack of 10
Product Code. 10177904
Brand: Thermo Scientific™ 290MA194
Only null left
Add to basket
Add to basket
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule Kit
Description 290 MA194
For Use With (Equipment) Thermo Scientific ISQ GC-MS Interface
Material SilTite Metal
Includes 2 SilTite Nuts, 10 Ferrules
Diameter (Metric) Inside 0.1 to 0.25 mm