Description
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
- SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
- Base of the ferrule is flat and forms a perfect seal with the MS interface
- The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
- Unlike other ferrules, do not need re-tightening after installation
Specifications
Specifications
| Type | Ferrule Kit |
| Description | 290 MT230 |
| For Use With (Equipment) | Thermo Scientific DSQ GC-MS Interface |
| Material | SilTite Metal |
| Includes | 2 SilTite Nuts, 10 Ferrules |
| Diameter (Metric) Inside | 0.32 mm |