Learn More
Cytiva Silicone Tubing/MicroPerpex
Product Code. 11390002 Shop All Cytiva Products
Change view
Click to view available options
Overall Length (Metric):
2 m
Unit Size:
Each
Product Code. Overall Length (Metric) unitSize
11390002 2 m Each
1 options
Product Code. 11390002
Brand: Cytiva 80106594
Only null left
Specifications
Type Tubing/MicroPerpex
Diameter (Metric) Outer 5 mm
Overall Length (Metric) 2 m
Material Silicone