Learn More
Masterflex™ Silicone Tubing
Click to view available options
Diameter (Metric) Inner:
0.8 mm
4.8 mm
Diameter (Metric) Outer:
4 mm
8 mm
Thickness (Metric):
0.8 mm
1.6 mm
Unit Size:
7.5m
Description
Use this tubing for pharmaceutical, biotechnology, food and beverage, aerospace and general lab applications. Saint Gobain Performance Plastics™ Silicone Tubing is peroxide-cured, offers excellent heat resistance and a low compression set.
- Odorless, nontoxic, translucent tubing
- Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements
- Manufactured according to GMP
- Sterilize by autoclave or gamma irradiation
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 0.8 mm |
| Material | Silicone |
| Diameter (Metric) Outer | 4 mm |
| Overall Length (Metric) | 7.6 m |
| Thickness (Metric) | 0.8 mm |
| Min. Operating Temperature (Metric) | -50°C |
| Color | Translucent |
| Max. Operating Temperature (Metric) | 230°c |