Learn More
Lenz Laborglasintrumente Silicone Sealing Rings
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01334532
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10060341
  • 2051.10 NOK / Pack of 10
Estimated Shipment: 25-06-2024
Description
  • For screw caps with bore
  • Material: silicone rubber (VMQ)
  • With bonded PTFE face
Specifications
Specifications
32 mm
Silicone