Learn More
DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer
Product Code. 15884598 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inside:
6 mm
8 mm
10 mm
12 mm
14 mm
16 mm
18 mm
20 mm
26 mm
32 mm
Joint Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Pack of 10
Product Code. Diameter (Metric) Inside Joint Size unitSize
15884598 12 mm GL 25 Pack of 10
15931541 6 mm GL 14 Pack of 10
10345191 6 mm GL 18 Each
15874598 10 mm GL 18 Pack of 10
15369797 8 mm GL 25 Pack of 10
10548411 26 mm GL 45 Each
15824618 14 mm GL 32 Pack of 10
10141831 8 mm GL 18 Each
15814618 12 mm GL 32 Pack of 10
10420303 10 mm GL 25 Each
15804618 10 mm GL 32 Pack of 10
15844618 18 mm GL 32 Pack of 10
15834618 16 mm GL 32 Pack of 10
15854618 20 mm GL 32 Pack of 10
15217458 32 mm GL 45 Pack of 10
15 options
Product Code. 15884598
Brand: DWK Life Sciences 286201607
Only null left
Add to basket
Add to basket
Seal for screw connection cap.
Suitable for PBT screw cap. Exemplary application: Fixing tubes
  • Suitable for PBT screw cap
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 12 mm
For Use With (Application) Fixing tubes; Sealing
Joint Size GL 25
Quantity 10/Pk.
Diameter (Metric) Outside 22 mm
Material Silicone