Learn More
DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer
Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Inside:
6 mm
8 mm
10 mm
12 mm
14 mm
16 mm
18 mm
20 mm
26 mm
32 mm
Joint Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Pack of 10
Description
Suitable for PBT screw cap. Exemplary application: Fixing tubes
- Suitable for PBT screw cap
Specifications
Specifications
| Diameter (Metric) Inside | 10 mm |
| For Use With (Application) | Fixing tubes; Sealing |
| Joint Size | GL 18 |
| Quantity | 10/Pk. |
| Diameter (Metric) Outside | 16 mm |
| Material | Silicone |