Learn More
DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer
Shop All DWK Life Sciences ProductsDescription
Suitable for PBT screw cap. Exemplary application: Fixing tubes
- Suitable for PBT screw cap
Specifications
Specifications
| Diameter (Metric) Inside | 10 mm |
| For Use With (Application) | Fixing tubes; Sealing |
| Joint Size | GL 25 |
| Quantity | 10/Pk. |
| Diameter (Metric) Outside | 22 mm |
| Material | Silicone |