Description
- O-ring design
- Made from FDA-acceptable grade silicone
- Suitable for use with both 304 and 316L stainless steel storage containers
- Wide range of sizes available
Specifications
Specifications
| For Use With (Application) | Leak-proof sealing |
| For Use With (Equipment) | 50L storage container |
| Material | Silicone |
| Type | Seal Ring |