Learn More
Lenz Laborglasintrumente Silicone Rubber Sealings, Septa
Product Code. p-7222602
Click to view available options
Diameter (Metric):
22 mm
29 mm
42 mm
Unit Size:
Pack of 100
Product Code. 10509791
Brand: Lenz Laborglasintrumente 01342932
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For piercing
  • For screw caps with bore
  • Material: silicone (VMQ)
Specifications
Material Silicone
Thickness (Metric) 2 mm
Diameter (Metric) 29 mm