Description
- Resistant to temperatures from –50 to +200°C (briefly up to +250°C)
- Deep frozen, hot or boiling substances and apparatus can be safely handled and positioned
- Sililcone rubber is extremely elastic and slip-proof; ideal for smooth surfaces
- Secure grip
Specifications
Specifications
| Material | Silicone Rubber |
| Color | White |
| For Use With (Application) | Protects Thumb and 1 Finger From Burns and Freezing |