Learn More
Sartorius™ Silicone O-ring
Product Code. 10517821 Shop All Sartorius Products
Change view
Click to view available options
Size:
40 x 2 mm
40 x 5 mm
42 x 3mm
45 x 3 mm
Unit Size:
Each
Product Code. Size unitSize
10517821 40 x 5 mm Each
10430873 45 x 3 mm Each
10337221 40 x 2 mm Each
10761553 42 x 3mm Each
4 options
Product Code. 10517821
Brand: Sartorius™ 6980110
Only null left
Specifications
For Use With (Equipment) Polycarbonate in-line filter holder
Quantity 1Each
Product Type O-Ring
Material Silicone
Size 40 x 5 mm