Learn More
Macherey-Nagel™ Silicone High Temperature Septa
Product Code. 10206501 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric) Outer:
13 mm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
10206501 13 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10206501 Supplier Macherey-Nagel™ Supplier No. 702623
  • ONLINE EXCLUSIVE PRICE
    975.00 NOK
    830.82 NOK / Pack of 50
    Save 144.18 NOK
    14% Off
Estimated Shipment: 10-12-2026
Specifications
Product Type High Temperature Septum
Diameter (Metric) Outer 13 mm
Hardness 60° Shore A
Temperature (Metric) Max 320°C
Color Red
Material Silicone
Quantity 50 Pack