Learn More
Hirschmann™ NS-ground Adapter, Silicone
Product Code. 11612229
Change view
Click to view available options
Thread Size:
A 45 to 29/32
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
11612229 A 45 to 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11612229 Supplier Hirschmann™ Supplier No. 9313742
  • ONLINE EXCLUSIVE PRICE
    507.15 NOK
    317.93 NOK / Pack of 1
    Save 189.22 NOK
    37% Off
Estimated Shipment: 18-12-2026
NS-ground adapter made of silicone for the adaption of bottle top mounted devices with GL45 to Pellet reservoir bottles.
  • Made from highly pure silicone
  • Adaptation on Pellet reservoir bottles
  • GL 45
  • NS29/32
Specifications
Product Type Adapter
Material Silicone
For Use With (Application) extraction reagents directly from a pulp bottle
For Use With (Equipment) Solarus titration
Thread Size A 45 to 29/32