Learn More
Saint Gobain Life Sciences Silicon Piston Ring
Click to view available options
For Use With (Equipment):
2 to 6 L Flask
50 to 500 mL Flask
500 to 2000 mL Flask
Unit Size:
Each
Pack of 5
Description
- Sturdy rubber Silicone piston rings for flasks
- For general lab applications
Specifications
Specifications
| Product Type | Piston Ring |
| Material | Silicon |
| For Use With (Equipment) | 2 to 6 L Flask |
| No. per Pack | 5 Pk. |
| Quantity | 5 Pk. |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto