missing translation for 'onlineSavingsMsg'
Learn More

Silicon(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. p-7051647
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11328858

Brand: Thermo Scientific Alfa Aesar 041711.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For vacuum deposition

Silicon(II) oxide can be used for synthesis of fine ceramic materials, such as silicon nitride, silicon carbide fine ceramic powder raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10097-28-6
Molecular Formula OSi
Molecular Weight (g/mol) 44.084
MDL Number MFCD00151536
InChI Key LIVNPJMFVYWSIS-UHFFFAOYSA-N
Synonym silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
PubChem CID 66241
ChEBI CHEBI:30588
IUPAC Name oxoniumylidynesilanide
SMILES [O+]#[Si-]

Specifications

Boiling Point 1880°C
Melting Point >1702°C
Physical Form Pieces
Quantity 100 g
Merck Index 14,8496
Solubility Information Insoluble in water.
Formula Weight 44.09
Percent Purity 99.9%
Odor Odorless
Density 2.13 g/mL
Assay (metals basis)
Chemical Name or Material Silicon(II) oxide, 6 to 15mm Pieces
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.