Learn More
Saint Gobain Life Sciences Silicon Folding Skirt Stopper
Product Code. 11714608
Change view
Click to view available options
Diameter (Metric):
7.1 mm
16 mm
Unit Size:
Pack of 100
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11714608 16 mm Pack of 20
11744598 7.1 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11714608 Supplier Saint Gobain Life Sciences Supplier No. 46701820
  • ONLINE EXCLUSIVE PRICE
    454.76 NOK
    318.44 NOK / Pack of 20
    Save 136.32 NOK
    29% Off
Estimated Shipment: 01-12-2026
Use this stopper for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Silicon Folding Skirt Stopper is particularly suited for all types of tissue culture work.
  • Two-part folding skirt stoppers
  • Designed with a double seal to keep container contents secure
  • Ideal for securing glass, metal and plastic containers
  • The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim
  • Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations
  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 16 mm
Quantity 1 Ea.
Color Transparent
Material Silicone
Product Type Folding Skirt Stopper