missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis excluding B), Thermo Scientific™

Product Code. 15443795
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
15443795 1 Ea. Each
1 options
This item is not returnable. View return policy

Product Code. 15443795

Brand: Thermo Scientific Alfa Aesar 041085.KS

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
MDL Number MFCD00049531
Formula Weight 40.1
Percent Purity 99.5%
Assay (metals basis excluding B)
Chemical Name or Material Silicon carbide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.