Learn More
Silicagel, Chem-Lab
Product Code. 11943315
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Polyethylene
Unit Size:
Each
Product Code. Quantity Packaging unitSize
11943315 1 kg Polyethylene Each
1 options
Product Code. 11943315
Brand: Chem-Lab CL00.1922.1000
Used as a desiccating agent in laboratory applications. Silicagel, Chem-Lab is a granular silicon dioxide compound that readily adsorbs water and protects processes and products from the effects of humidity and moisture.
Silica gel's high specific surface area allows it to adsorb water readily, making it useful as a desiccant (drying agent). In chemistry, silica gel is used in chromatography as a stationary phase.
Specifications
Physical Form Solid
CAS 7631-86-9
Density 0.75g/cm3
Material Silicon Dioxide
Quantity 1 kg
Molecular Formula O2Si
Loss on Drying <2%
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Packaging Polyethylene
Solubility Information Insoluble
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name dioxosilane
Molecular Weight (g/mol) 60.08
PubChem CID 24261
ChEBI CHEBI:30563
Formula Weight 60.08g/mol
Water Absorption Max capacity: >25%
Chemical Name or Material Silicon Dioxide