Learn More
Palintest™ Silica Test Kit
Product Code. 11998047
Change view
Click to view available options
Quantity:
200 Pack
Unit Size:
Pack of 200
Product Code. Quantity unitSize
11998047 200 Pack Pack of 200
1 options
Product Code. 11998047
Brand: Palintest™ AP181
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Concentration 0 to 4 mg/L
Quantity 200 Pack
compliance-icons compliance-icons
Product Identifier
  • X200 REFILL PHOTOM SILICA 0-4PPM
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : boric acid, sulphamic acid, ammonium molybdate, citric acid,adipic acid