missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Silica Test Disc

Product Code. 11968896
Change view
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11968896 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11968896 Supplier Palintest™ Supplier No. CM181

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Palintest™ comparator

The Palintest™ comparator with interchangeable color discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.

Specifications

Concentration 0 to 4 mg/L
Packaging 50/Pk.
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.