Learn More
Silica Standard Solution, WTW™
Catalog Number 11743976
Click to view available options
Quantity:
500 mL
Physical Form Liquid
Concentration 1000mg/L
Solution Contains Silica
Traceability to NIST Yes
pH 13.5
Boiling Point 100°C
Color Colorless
Quantity 500 mL
Solubility Soluble in water
compliance-icons
Product Identifier
  • Silicon standard solution traceable to SRM from NIST SiO
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.