Learn More
Macherey-Nagel™ Silica Glass Plates
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
10 x 20 cm
Unit Size:
Pack of 50
Description
HPTLC glass plates in different plate sizes for many different applications. Silica gel (SiOH) impregnated with caffeine for the analysis of polycyclic aromatic hydrocarbons.
Specifications
Specifications
| Material | Silica 60 |
| Particle Size | 2 to 10 μm |
| Pore Size | 60 Å |
| Quantity | 50 Pk. |
| Type | Glass Plate |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.20 mm |
| Dimensions (L x W) | 10 x 20 cm |