Learn More
Silica gel orange, Honeywell™
Granulate (0.2-1 mm)
Brand: Honeywell Research Chemicals 10087H-1KG
Additional Details : CAS Number : 7783-83-7 Weight : 1.00000kg
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Beads | |
≥90.00% | |
1 kg | |
Max. 2 % |
7631-86-9,7783-83-7 | |
≤100.00% | |
Silica gel | |
Plastic Bottle |